2-(4-Chlorophenyl)propionic acid 96%
Lieferant: Apollo Scientific
OR951001-1G
OR951001-25G
OR951001-5G
APOSOR951001-1GEA
132
EUR
APOSOR951001-1G
APOSOR951001-25G
APOSOR951001-5G
2-(4-Chlorophenyl)propionic acid 96%
2-(4-Chlorophenyl)propionic acid
Formel:
C₉H₉ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044670 CAS-Nummer: 938-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...