2-(4-Chlorophenyl)propionic acid 96%

Lieferant: Apollo Scientific

OR951001-1G OR951001-25G OR951001-5G
APOSOR951001-1GEA 132 EUR
APOSOR951001-1G APOSOR951001-25G APOSOR951001-5G
2-(4-Chlorophenyl)propionic acid 96%
2-(4-Chlorophenyl)propionic acid
Formel: C₉H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00044670
CAS-Nummer: 938-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR