Methyl 4-pyridylacetate hydrochloride 95%

Lieferant: Apollo Scientific

OR951032-1G OR951032-25G OR951032-5G
APOSOR951032-1GEA 161 EUR
APOSOR951032-1G APOSOR951032-25G APOSOR951032-5G
Methyl 4-pyridylacetate hydrochloride 95%
Methyl 4-pyridylacetate hydrochloride
Formel: C₈H₉NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00236622
CAS-Nummer: 70199-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR