3-Methanesulphonyl-phenol 97%

Lieferant: Apollo Scientific

OR951080-1G OR951080-250MG
APOSOR951080-1GEA 580 EUR
APOSOR951080-1G APOSOR951080-250MG
3-Methanesulphonyl-phenol 97%
Formel: C₇H₈O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD11839805
CAS-Nummer: 14763-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR