4-Methylpiperidine-4-carbonitrile hydrochloride 95%

Lieferant: Apollo Scientific

OR951098-1G
APOSOR951098-1GEA 148 EUR
APOSOR951098-1G
4-Methylpiperidine-4-carbonitrile hydrochloride 95%
4-Methylpiperidine-4-carbonitrile hydrochloride
Formel: C₇H₁₂N₂·ClH
Molecular Weight: 160,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406584
CAS-Nummer: 948894-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR