Methyl-4-oxocyclohexanecarboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-oxocyclohexanecarboxylate, 4-Oxo-cyclohexanecarboxylic acid methyl ester
OR951132-100G
APOSOR951132-100GEA
219
EUR
APOSOR951132-100G
Methyl-4-oxocyclohexanecarboxylate 95%
Methyl-4-oxocyclohexanecarboxylate
Formel:
C₈H₁₂O₃ Molecular Weight: 156.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270324 CAS-Nummer: 6297-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...