2-(4-Methylphenoxy)ethylamine 95%

Lieferant: Apollo Scientific

Synonyme: [2-(4-Methylphenoxy)ethyl]amine, 2-(p-Tolyloxy)ethanamine

OR951144-1G
APOSOR951144-1GEA 78.3 EUR
APOSOR951144-1G
2-(4-Methylphenoxy)ethylamine 95%
2-(4-Methylphenoxy)ethylamine
Formel: C₉H₁₃NO
Molecular Weight: 151,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02598961
CAS-Nummer: 26583-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR