2-Hydroxy-5-chloromethylbenzaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 5-(Chloromethyl)-2-hydroxybenzaldehyde

OR951182-1G
APOSOR951182-1GEA 72.5 EUR
APOSOR951182-1G
2-Hydroxy-5-chloromethylbenzaldehyde 95%
2-Hydroxy-5-chloromethylbenzaldehyde
Formel: C₈H₇ClO₂
Molecular Weight: 170.593 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00226977
CAS-Nummer: 23731-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR