1-(3,4-Dichlorophenyl)ethanol 95%

Lieferant: Apollo Scientific

OR951245-25G
APOSOR951245-25GEA 213 EUR
APOSOR951245-25G
1-(3,4-Dichlorophenyl)ethanol 95%
1-(3,4-Dichlorophenyl)ethanol
Formel: C₈H₈Cl₂O
Molecular Weight: 191.055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021865
CAS-Nummer: 1475-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR