4-Methylpyrazole-3-carboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 4-Methylpyrazole-3-carboxylic Acid

OR951257-5G
APOSOR951257-5GEA 171 EUR
APOSOR951257-5G
4-Methylpyrazole-3-carboxylic acid 98%
4-Methylpyrazole-3-carboxylic acid
Formel: C₅H₆N₂O₂
Molecular Weight: 126.113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16495896
CAS-Nummer: 82231-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR