2-(2-Chlorphenyl)ethanol 98%

Lieferant: Apollo Scientific

OR951312-100G OR951312-25G
APOSOR951312-100GEA 306 EUR
APOSOR951312-100G APOSOR951312-25G
2-(2-Chlorphenyl)ethanol 98%
2-(2-Chlorphenyl)ethanol
Formel: C₈H₉ClO
Molecular Weight: 156.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002888
CAS-Nummer: 19819-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR