2-Cyanbenzoesäure 96%

Lieferant: Apollo Scientific
Achtung

OR951322-25G
APOSOR951322-25GEA 183 EUR
APOSOR951322-25G
2-Cyanbenzoesäure 96%
2-Cyanbenzoesäure
Formel: C₈H₅NO₂
Molecular Weight: 147.13 g/mol
Schmelzpunkt: 212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045796
CAS-Nummer: 3839-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR