3-(1-Hydroxyethyl)phenol 98%

Lieferant: Apollo Scientific

OR951348-25G OR951348-5G
APOSOR951348-25GEA 127 EUR
APOSOR951348-25G APOSOR951348-5G
3-(1-Hydroxyethyl)phenol 98%
3-(1-Hydroxyethyl)phenol
Formel: C₈H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00238621
CAS-Nummer: 2415-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR