o,α-Dimethylbenzylalkohol 95%

Lieferant: Apollo Scientific

Synonyme: 2-(1-Hydroxyethyl)toluene, 2-Methylphenyl methyl carbinol, α,2-Dimethylbenzyl alcohol, 1-(2-Methylphenyl)ethanol, 1-(o-Tolyl)ethanol

OR951356-25G
APOSOR951356-25GEA 101 EUR
APOSOR951356-25G
o,α-Dimethylbenzylalkohol 95%
o,α-Dimethylbenzylalkohol
Formel: C₉H₁₂O
Molecular Weight: 136.19
Lagertemperatur: Raumtemperatur
MDL: MFCD00046632
CAS-Nummer: 7287-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR