5-(Hydroxymethyl)quinolin-8-ol 95%

Lieferant: Apollo Scientific

OR951368-1G
APOSOR951368-1GEA 294 EUR
APOSOR951368-1G
5-(Hydroxymethyl)quinolin-8-ol 95%
5-(Hydroxymethyl)quinolin-8-ol
Formel: C₁₀H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03614757
CAS-Nummer: 4053-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR