Pseudopelletierine oxime 98%

Lieferant: Apollo Scientific

OR951413-100G
APOSOR951413-100GEA 231 EUR
APOSOR951413-100G
Pseudopelletierine oxime 98%
Pseudopelletierine oxime
Formel: C₉H₁₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03755545
CAS-Nummer: 6164-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR