3,4-Dihydro-1H-quinoxalin-2-one 95%

Lieferant: Apollo Scientific

OR951424-1G OR951424-5G
APOSOR951424-1GEA 55.1 EUR
APOSOR951424-1G APOSOR951424-5G
3,4-Dihydro-1H-quinoxalin-2-one 95%
3,4-Dihydro-1H-quinoxalin-2-one
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00204133
CAS-Nummer: 59564-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR