3-Methylquinazoline-2,4(1H,3H)-dione 95%

Lieferant: Apollo Scientific

OR951434-1G
APOSOR951434-1GEA 632 EUR
APOSOR951434-1G
3-Methylquinazoline-2,4(1H,3H)-dione 95%
3-Methylquinazoline-2,4(1H,3H)-dione
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01534101
CAS-Nummer: 607-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR