5-Chloro-2-furoic acid 98%

Lieferant: Apollo Scientific

Synonyme: 5-Chlorofuran-2-carboxylic acid

OR951451-5G
APOSOR951451-5GEA 58 EUR
APOSOR951451-5G
5-Chloro-2-furoic acid 98%
5-Chloro-2-furoic acid
Formel: C₅H₃ClO₃
Molecular Weight: 146.529 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093272
CAS-Nummer: 618-30-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR