4-(Methylsulphonyl)benzyl alcohol 98%
Lieferant: Apollo Scientific
Synonyme:
4-(Methylsulphonyl)benzyl Alcohol
OR951461-25G
APOSOR951461-25GEA
71.2
EUR
APOSOR951461-25G
4-(Methylsulphonyl)benzyl alcohol 98%
4-(Methylsulphonyl)benzyl alcohol
Formel:
C₈H₁₀O₃S Molecular Weight: 186.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425695 CAS-Nummer: 22821-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...