5,6,7,8-Tetrahydroquinazolin-2-amine 95%

Lieferant: Apollo Scientific

OR951480-1G
APOSOR951480-1GEA 75.4 EUR
APOSOR951480-1G
5,6,7,8-Tetrahydroquinazolin-2-amine 95%
5,6,7,8-Tetrahydroquinazolin-2-amine
Formel: C₈H₁₁N₃
Molecular Weight: 149.193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08060043
CAS-Nummer: 2305-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR