1-(3,4-Dichlorophenyl)ethylamine 97%

Lieferant: Apollo Scientific

Synonyme: 1-(3,4-Dichlorophenyl)ethanamine

OR951494-5G
APOSOR951494-5GEA 114 EUR
APOSOR951494-5G
1-(3,4-Dichlorophenyl)ethylamine 97%
1-(3,4-Dichlorophenyl)ethylamine
Formel: C₈H₉Cl₂N
Molecular Weight: 190.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01123279
CAS-Nummer: 74877-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR