4-(Aminomethyl)-N,N-dimethylbenzamide 98%

Lieferant: Apollo Scientific

OR951509-1G OR951509-250MG
APOSOR951509-1GEA 415 EUR
APOSOR951509-1G APOSOR951509-250MG
4-(Aminomethyl)-N,N-dimethylbenzamide 98%
4-(Aminomethyl)-N,N-dimethylbenzamide
Formel: C₁₀H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07186282
CAS-Nummer: 164648-76-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR