3,6-Dimethyl-2-nitrophenol 95%
Lieferant: Apollo Scientific
OR951519-1G
APOSOR951519-1GEA
261
EUR
APOSOR951519-1G
3,6-Dimethyl-2-nitrophenol 95%
3,6-Dimethyl-2-nitrophenol
Formel:
C₈H₉NO₃ Molecular Weight: 167,162 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12198862 CAS-Nummer: 71608-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...