5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine

OR951539-250MG
APOSOR951539-250MGEA 112 EUR
APOSOR951539-250MG
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Formel: C₆H₄Cl₂N₄
Molecular Weight: 203,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18642394
CAS-Nummer: 1124383-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR