1-Hydroxycyclohexanecarboxylic acid 95%

Lieferant: Apollo Scientific

OR951583-5G
APOSOR951583-5GEA 126 EUR
APOSOR951583-5G
1-Hydroxycyclohexanecarboxylic acid 95%
1-Hydroxycyclohexanecarboxylic acid
Formel: C₇H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00045477
CAS-Nummer: 1123-28-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR