5-(Methylamino)nicotinic acid 95%

Lieferant: Apollo Scientific

OR951641-1G OR951641-250MG
APOSOR951641-1GEA 311 EUR
APOSOR951641-1G APOSOR951641-250MG
5-(Methylamino)nicotinic acid 95%
5-(Methylamino)nicotinic acid
Formel: C₇H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04971939
CAS-Nummer: 91702-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR