Methyl-3-aminopicolinat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-aminopyridine-2-carboxylate, Methyl 3-aminopicolinate

OR951644-25G
APOSOR951644-25GEA 381 EUR
APOSOR951644-25G
Methyl-3-aminopicolinat 98%
Methyl-3-aminopicolinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08695602
CAS-Nummer: 36052-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR