3-Methyl-2-oxo-butanoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-2-oxobutanoic acid

OR951671-1G
APOSOR951671-1GEA 72.5 EUR
APOSOR951671-1G
3-Methyl-2-oxo-butanoic acid 95%
3-Methyl-2-oxo-butanoic acid
Formel: C₅H₈O₃
Molecular Weight: 116.115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040427
CAS-Nummer: 759-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR