5-Formyl-2-methoxybenzoic acid 96%

Lieferant: Apollo Scientific

OR951686-25G OR951686-5G
APOSOR951686-25GEA 590 EUR
APOSOR951686-25G APOSOR951686-5G
5-Formyl-2-methoxybenzoic acid 96%
5-Formyl-2-methoxybenzoic acid
Formel: C₉H₈O₄
Molecular Weight: 180.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01442588
CAS-Nummer: 84923-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR