4-Chloro-1H-indole-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR951731-1G OR951731-5G
APOSOR951731-1GEA 46.4 EUR
APOSOR951731-1G APOSOR951731-5G
4-Chloro-1H-indole-2-carboxylic acid 95%
4-Chloro-1H-indole-2-carboxylic acid
Formel: C₉H₆ClNO₂
Molecular Weight: 195.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664470
CAS-Nummer: 24621-73-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR