2-(2-Aminoethoxy)benzonitril 95%
Lieferant: Apollo Scientific
OR951747-1G
OR951747-250MG
APOSOR951747-1GEA
364
EUR
APOSOR951747-1G
APOSOR951747-250MG
2-(2-Aminoethoxy)benzonitril 95%
2-(2-Aminoethoxy)benzonitril
Formel:
C₉H₁₀N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD16495889 CAS-Nummer: 57276-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...