2-(2-Aminoethoxy)benzonitril 95%

Lieferant: Apollo Scientific

OR951747-1G OR951747-250MG
APOSOR951747-1GEA 364 EUR
APOSOR951747-1G APOSOR951747-250MG
2-(2-Aminoethoxy)benzonitril 95%
2-(2-Aminoethoxy)benzonitril
Formel: C₉H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD16495889
CAS-Nummer: 57276-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR