5-Chloroimidazo[1,2-a]pyrazine 96%

Lieferant: Apollo Scientific

OR951845-1G OR951845-250MG
APOSOR951845-1GEA 451 EUR
APOSOR951845-1G APOSOR951845-250MG
5-Chloroimidazo[1,2-a]pyrazine 96%
5-Chloroimidazo[1,2-a]pyrazine
Formel: C₆H₄ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13193447
CAS-Nummer: 63744-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR