N-Methoxy-N-methylcyclopent-3-ene-1-carboxamide 95%

Lieferant: Apollo Scientific

OR951850-1G
APOSOR951850-1GEA 560 EUR
APOSOR951850-1G
N-Methoxy-N-methylcyclopent-3-ene-1-carboxamide 95%
N-Methoxy-N-methylcyclopent-3-ene-1-carboxamide
Formel: C₈H₁₃NO₂
Molecular Weight: 155,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07370045
CAS-Nummer: 248275-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR