2-[4-(Methylthio)phenoxy]ethylamine 95%

Lieferant: Apollo Scientific

OR951874-1G OR951874-250MG
APOSOR951874-1GEA 364 EUR
APOSOR951874-1G APOSOR951874-250MG
2-[4-(Methylthio)phenoxy]ethylamine 95%
2-[4-(Methylthio)phenoxy]ethylamine
Formel: C₉H₁₃NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD13370835
CAS-Nummer: 1203188-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR