Methyl 5-formylthiophene-2-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-formylthiophene-2-carboxylate, 5-Formylthiophene-2-carboxylic acid methyl ester

OR951877-25G
APOSOR951877-25GEA 594 EUR
APOSOR951877-25G
Methyl 5-formylthiophene-2-carboxylate 98%
Methyl-5-formylthiophene-2-carboxylate
Formel: C₇H₆O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD01859942
CAS-Nummer: 67808-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR