3'-Methylpropiophenon 96%

Lieferant: Apollo Scientific

OR951894-25G
APOSOR951894-25GEA 139 EUR
APOSOR951894-25G
3'-Methylpropiophenon 96%
3'-Methylpropiophenon
Formel: C₁₀H₁₂O
Molecular Weight: 148.2017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00027014
CAS-Nummer: 51772-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR