5-Methyl-2-phenyl-1H-imidazole-4-methanol 97%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxymethyl-5-methyl-2-phenylimidazole

OR951906-1G
APOSOR951906-1GEA 74.1 EUR
APOSOR951906-1G
5-Methyl-2-phenyl-1H-imidazole-4-methanol 97%
5-Methyl-2-phenyl-1H-imidazole-4-methanol
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188.226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047016
CAS-Nummer: 13682-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR