Methyl-2-benzo[b]thiophencarboxylat 95%

Lieferant: Apollo Scientific

OR951927-1G OR951927-5G
APOSOR951927-1GEA 0 EUR
APOSOR951927-1G APOSOR951927-5G
Methyl-2-benzo[b]thiophencarboxylat 95%
Methyl-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₈O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00067791
CAS-Nummer: 22913-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR