3-Chloro-5-methoxypyridine 98%

Lieferant: Apollo Scientific

OR952205-1G
APOSOR952205-1GEA 58 EUR
APOSOR952205-1G
3-Chloro-5-methoxypyridine 98%
3-Chloro-5-methoxypyridine
Formel: C₆H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09910244
CAS-Nummer: 95881-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR