Ethyl 5-methylpyrazine-2-carboxylate 95%

Lieferant: Apollo Scientific

OR952290-25G
APOSOR952290-25GEA 125 EUR
APOSOR952290-25G
Ethyl 5-methylpyrazine-2-carboxylate 95%
Ethyl 5-methylpyrazine-2-carboxylate
Formel: C₈H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05662506
CAS-Nummer: 41110-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR