3-Methyl-3H-imidazo[4,5-b]pyridine 98%

Lieferant: Apollo Scientific

OR952327-1G
APOSOR952327-1GEA 208 EUR
APOSOR952327-1G
3-Methyl-3H-imidazo[4,5-b]pyridine 98%
3-Methyl-3H-imidazo[4,5-b]pyridine
Formel: C₇H₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03676333
CAS-Nummer: 6688-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR