6-Ethyl-4-methoxy-2-pyranone 97%

Lieferant: Apollo Scientific

OR952337-1G
APOSOR952337-1GEA 128 EUR
APOSOR952337-1G
6-Ethyl-4-methoxy-2-pyranone 97%
6-Ethyl-4-methoxy-2-pyranone
Formel: C₈H₁₀O₃
Molecular Weight: 154,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09991688
CAS-Nummer: 106950-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR