4-(Tetrahydrofuran-2-ylmethoxy)benzoic acid 98%

Lieferant: Apollo Scientific
Gefahr

OR952349-250MG
APOSOR952349-250MGEA 190 EUR
APOSOR952349-250MG
4-(Tetrahydrofuran-2-ylmethoxy)benzoic acid 98%
4-(Tetrahydrofuran-2-ylmethoxy)benzoic acid
Formel: C₁₂H₁₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03970016
CAS-Nummer: 565194-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR