4'-(1-Pyrazolyl)acetophenon 98%
Lieferant: Apollo Scientific
OR952366-10G
OR952366-25G
OR952366-5G
APOSOR952366-10GEA
0
EUR
APOSOR952366-10G
APOSOR952366-25G
APOSOR952366-5G
4'-(1-Pyrazolyl)acetophenon 98%
4'-(1-Pyrazolyl)acetophenon
Formel:
C₁₁H₁₀N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD08572143 CAS-Nummer: 25699-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...