4'-(1-Pyrazolyl)acetophenon 98%

Lieferant: Apollo Scientific

OR952366-10G OR952366-25G OR952366-5G
APOSOR952366-10GEA 0 EUR
APOSOR952366-10G APOSOR952366-25G APOSOR952366-5G
4'-(1-Pyrazolyl)acetophenon 98%
4'-(1-Pyrazolyl)acetophenon
Formel: C₁₁H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08572143
CAS-Nummer: 25699-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR