6-Methoxy-1H-pyrrolo[2,3-b]pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 6-Methoxy-7-azaindole

OR952412-1G
APOSOR952412-1GEA 126 EUR
APOSOR952412-1G
6-Methoxy-1H-pyrrolo[2,3-b]pyridine 95%
6-Methoxy-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₈N₂O
Molecular Weight: 148,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659665
CAS-Nummer: 896722-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR