N-Ethyl-3,4-dimethylbenzylamine 95%

Lieferant: Apollo Scientific

OR952456-1G OR952456-5G
APOSOR952456-1GEA 226 EUR
APOSOR952456-1G APOSOR952456-5G
N-Ethyl-3,4-dimethylbenzylamine 95%
N-Ethyl-3,4-dimethylbenzylamine
Formel: C₁₁H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD11204729
CAS-Nummer: 39180-83-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR