N-Ethyl-3,4-dimethylbenzylamine 95%
Lieferant: Apollo Scientific
OR952456-1G
OR952456-5G
APOSOR952456-1GEA
226
EUR
APOSOR952456-1G
APOSOR952456-5G
N-Ethyl-3,4-dimethylbenzylamine 95%
N-Ethyl-3,4-dimethylbenzylamine
Formel:
C₁₁H₁₇N Lagertemperatur: Raumtemperatur |
MDL:
MFCD11204729 CAS-Nummer: 39180-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...